Assay Details
Gene Symbol: Eif4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191048.2 | NP_001177977.2 | 134 | |||
GenBank mRNA | HP496780.1 | - | 134 | |||
HP556264.1 | - | 134 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Eif4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191048.2 | NP_001177977.2 | 134 | |||
GenBank mRNA | HP496780.1 | - | 134 | |||
HP556264.1 | - | 134 |