Assay Details
Gene Symbol: LOC408824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP501815.1 | - | 64 | |||
HP516642.1 | - | 64 | ||||
HP516643.1 | - | 64 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC408824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP501815.1 | - | 64 | |||
HP516642.1 | - | 64 | ||||
HP516643.1 | - | 64 |