Assay Details
Gene Symbol: LOC100576381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP502158.1 | - | 87 | |||
HP516001.1 | - | 87 | ||||
HP516002.1 | - | 87 | ||||
HP565565.1 | - | 87 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100576381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP502158.1 | - | 87 | |||
HP516001.1 | - | 87 | ||||
HP516002.1 | - | 87 | ||||
HP565565.1 | - | 87 |