Assay Details
Gene Symbol: Vamp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242527.1 | NP_001229456.1 | 160 | |||
GenBank mRNA | HP517820.1 | - | 160 | |||
HP517821.1 | - | 160 | ||||
HP574848.1 | - | 160 |
