Assay Details
Gene Symbol: LOC726798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP523160.1 | - | 91 | |||
HP537373.1 | - | 91 | ||||
HP576616.1 | - | 91 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC726798 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP523160.1 | - | 91 | |||
HP537373.1 | - | 91 | ||||
HP576616.1 | - | 91 |