Assay Details
Gene Symbol: Dnmt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171051.2 | NP_001164522.2 | 149 | |||
GenBank mRNA | HP525775.1 | - | 148 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnmt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171051.2 | NP_001164522.2 | 149 | |||
GenBank mRNA | HP525775.1 | - | 148 |