Assay Details
Gene Symbol: PDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242513.1 | NP_001229442.1 | 69 | |||
GenBank mRNA | HP537189.1 | - | 69 | |||
HP549405.1 | - | 69 | ||||
HP573896.1 | - | 69 |
