Assay Details
Gene Symbol: LOC551787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP493741.1 | - | 119 | |||
HP539692.1 | - | 119 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC551787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP493741.1 | - | 119 | |||
HP539692.1 | - | 119 |