Assay Details
Gene Symbol: LOC411607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP563125.1 | - | 92 | |||
KX898002.1 | - | 92 |
Gene Symbol: LOC725566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP469536.1 | - | 92 |
