Assay Details
Gene Symbol: Gycbeta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011632.2 | NP_001011632.1 | 64 | |||
GenBank mRNA | AB181489.1 | - | 64 | |||
AY769960.1 | - | 64 | ||||
HP493417.1 | - | 64 |
