Assay Details
Gene Symbol: Gycbeta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB181489.1 | - | 78 | |||
HP493417.1 | - | 78 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gycbeta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB181489.1 | - | 78 | |||
HP493417.1 | - | 78 |