Assay Details
Gene Symbol: AChE-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040230.1 | NP_001035320.1 | 103 | |||
GenBank mRNA | AB181702.1 | - | 103 | |||
AF213012.1 | - | 103 | ||||
HP496809.1 | - | 103 | ||||
HP573537.1 | - | 103 | ||||
KU532289.1 | - | 103 |
