Assay Details
Gene Symbol: sGC-alpha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011650.1 | NP_001011650.1 | 107 | |||
GenBank mRNA | AB193550.1 | - | 107 | |||
HP499071.1 | - | 107 | ||||
HP539807.1 | - | 107 |
