Assay Details
Gene Symbol: Ho | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011641.1 | NP_001011641.1 | 181 | |||
GenBank mRNA | AB194707.1 | - | 181 | |||
HP479252.1 | - | 181 | ||||
HP520551.1 | - | 181 |
