Assay Details
Gene Symbol: Abscam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040235.2 | NP_001035325.2 | 142 | |||
GenBank mRNA | AB257298.1 | - | 142 | |||
AB269871.1 | - | 142 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abscam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040235.2 | NP_001035325.2 | 142 | |||
GenBank mRNA | AB257298.1 | - | 142 | |||
AB269871.1 | - | 142 |