Assay Details
Gene Symbol: IF-2mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011586.1 | NP_001011586.1 | 103 | |||
GenBank mRNA | AF069739.1 | - | 103 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IF-2mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011586.1 | NP_001011586.1 | 103 | |||
GenBank mRNA | AF069739.1 | - | 103 |