Assay Details
Gene Symbol: RpLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185144.1 | NP_001172073.1 | 109 | |||
GenBank mRNA | AF134816.1 | - | 109 | |||
DB743243.1 | - | 109 | ||||
DB777493.1 | - | 109 | ||||
HP498692.1 | - | 109 |
