Assay Details
Gene Symbol: Obp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011590.1 | NP_001011590.1 | 95 | |||
GenBank mRNA | AF166496.1 | - | 95 | |||
AF393494.1 | - | 95 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Obp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011590.1 | NP_001011590.1 | 95 | |||
GenBank mRNA | AF166496.1 | - | 95 | |||
AF393494.1 | - | 95 |