Assay Details
Gene Symbol: Obp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011589.1 | NP_001011589.1 | 71 | |||
GenBank mRNA | AF393492.1 | - | 71 | |||
AF393495.1 | - | 71 | ||||
HP469449.1 | - | 71 |
