Assay Details
Gene Symbol: CSP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011583.2 | NP_001011583.1 | 75 | |||
GenBank mRNA | AF481963.1 | - | 75 | |||
HP518052.1 | - | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001011583.2 transcripts.
