Assay Details
Gene Symbol: CSP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011583.2 | NP_001011583.1 | 87 | |||
GenBank mRNA | AF481963.1 | - | 87 | |||
AJ973401.1 | - | 87 | ||||
DQ855484.1 | - | 87 | ||||
HP518052.1 | - | 87 |
