Assay Details
Gene Symbol: LOC409927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP495981.1 | - | 89 | |||
HP564909.1 | - | 89 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC409927 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP495981.1 | - | 89 | |||
HP564909.1 | - | 89 |