Assay Details
Gene Symbol: NFRKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040237.1 | NP_001035327.1 | 160 | |||
GenBank mRNA | HP505106.1 | - | 160 | |||
HP531869.1 | - | 160 | ||||
HP531870.1 | - | 160 |
