Assay Details
Gene Symbol: LOC406105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134947.1 | NP_001128419.1 | 138 | |||
GenBank mRNA | HP464232.1 | - | 138 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC406105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134947.1 | NP_001128419.1 | 138 | |||
GenBank mRNA | HP464232.1 | - | 138 |