Assay Details
Gene Symbol: Hsp90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160064.1 | NP_001153536.1 | 144 | |||
GenBank mRNA | FJ713701.1 | - | 144 | |||
FJ713702.1 | - | 144 | ||||
FJ713706.1 | - | 144 | ||||
FJ713707.1 | - | 144 | ||||
HP470852.1 | - | 144 | ||||
HP512306.1 | - | 144 |
