Assay Details
Gene Symbol: Pet117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164809.1 | NP_001158281.1 | 104 | |||
GenBank mRNA | DB735046.1 | - | 104 | |||
DB748789.1 | - | 104 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pet117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164809.1 | NP_001158281.1 | 104 | |||
GenBank mRNA | DB735046.1 | - | 104 | |||
DB748789.1 | - | 104 |