Assay Details
Gene Symbol: mRpL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185104.1 | NP_001172033.1 | 120 | |||
GenBank mRNA | BI514960.1 | - | 120 | |||
HP492393.1 | - | 120 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: mRpL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185104.1 | NP_001172033.1 | 120 | |||
GenBank mRNA | BI514960.1 | - | 120 | |||
HP492393.1 | - | 120 |