Assay Details
Gene Symbol: Ube2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242519.1 | NP_001229448.1 | 82 | |||
GenBank mRNA | HP478084.1 | - | 82 | |||
HP545157.1 | - | 82 | ||||
HP575740.1 | - | 82 |
