Assay Details
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242544.1 | NP_001229473.1 | 103 | |||
GenBank mRNA | HP466171.1 | - | 103 | |||
HP499643.1 | - | 103 | ||||
HP518894.1 | - | 103 | ||||
HP547809.1 | - | 103 |
