Assay Details
Gene Symbol: ATP5G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242656.1 | NP_001229585.1 | 116 | |||
NM_001242657.1 | NP_001229586.1 | 116 | ||||
GenBank mRNA | DB737491.1 | - | 116 | |||
HP461329.1 | - | 116 | ||||
HP461330.1 | - | 116 | ||||
HP512162.1 | - | 116 | ||||
HP565942.1 | - | 116 | ||||
HP565943.1 | - | 116 |
