Assay Details
Gene Symbol: LPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101377.5 | NP_172961.1 | 6 - 7 | 977 | 123 | |
GenBank mRNA | AB053950.1 | - | 5 - 6 | 695 | 123 | |
AB061407.1 | - | 6 - 7 | 898 | 123 | ||
AY087673.1 | - | 6 - 7 | 890 | 123 | ||
BT003912.1 | - | 6 - 7 | 890 | 123 | ||
BT004969.1 | - | 5 - 6 | 695 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
