Assay Details
Gene Symbol: MYB58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101514.3 | NP_173098.1 | 2 - 3 | 517 | 71 | |
GenBank mRNA | AB493459.1 | - | 2 - 3 | 273 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
