Assay Details
Gene Symbol: DMS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101768.4 | NP_173344.2 | 11 - 12 | 1521 | 79 | |
GenBank mRNA | AK221449.1 | - | 11 - 12 | 1474 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
