Assay Details
Gene Symbol: SOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126259.4 | NP_178307.2 | 13 - 14 | 1340 | 124 | |
GenBank mRNA | AY062746.1 | - | 13 - 14 | 1332 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
