Assay Details
Gene Symbol: AT3G01950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111061.4 | NP_186844.1 | 1 | 200 | 75 | |
GenBank mRNA | AK119150.1 | - | 1 | 157 | 75 | |
BT004658.1 | - | 1 | 108 | 75 |
Gene Symbol: AT3G01960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111062.2 | NP_186845.1 | 2 | 1124 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
