Assay Details
Gene Symbol: LPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111128.2 | NP_566177.1 | 1 - 2 | 398 | 100 | |
GenBank mRNA | AY084915.1 | - | 1 - 2 | 231 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
