Assay Details
Gene Symbol: LPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084626.2 | NP_001078095.1 | 7 - 8 | 1162 | 68 | |
NM_001084627.1 | NP_001078096.1 | 7 - 8 | 875 | 68 | ||
NM_001084628.2 | NP_001078097.1 | 8 - 9 | 1499 | 68 | ||
NM_001084629.2 | NP_001078098.1 | 7 - 8 | 987 | 68 | ||
NM_001337419.1 | NP_001327801.1 | 8 - 9 | 1980 | 68 | ||
NM_001337420.1 | NP_001327802.1 | 6 - 7 | 2489 | 68 | ||
NM_001337421.1 | NP_001319453.1 | 8 - 9 | 1066 | 68 | ||
NM_001337422.1 | NP_001327800.1 | 7 - 8 | 1425 | 68 | ||
NM_001337423.1 | NP_001327797.1 | 8 - 9 | 1305 | 68 | ||
NM_001337424.1 | NP_001327798.1 | 7 - 8 | 2381 | 68 | ||
NM_001337425.1 | NP_001327799.1 | 7 - 8 | 2100 | 68 | ||
NM_111128.2 | NP_566177.1 | 8 - 9 | 1255 | 68 | ||
GenBank mRNA | AY084915.1 | - | 8 - 9 | 1088 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
