Assay Details
Gene Symbol: SQN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127141.3 | NP_565381.1 | 3 - 4 | 913 | 72 | |
GenBank mRNA | AY026065.1 | - | 3 - 4 | 913 | 72 | |
BT028999.1 | - | 3 - 4 | 758 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
