Assay Details
Gene Symbol: GSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125685.1 | NP_001119157.1 | 2 - 3 | 235 | 92 | |
NM_001342670.1 | NP_001318461.1 | 3 - 4 | 267 | 92 | ||
NM_120356.4 | NP_195898.2 | 2 - 3 | 260 | 92 | ||
GenBank mRNA | BT010718.1 | - | 2 - 3 | 235 | 92 | |
BT012424.1 | - | 2 - 3 | 154 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
