Assay Details
Gene Symbol: NDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084449.2 | NP_001077918.1 | 4 - 5 | 560 | 71 | |
NM_001335640.1 | NP_001323956.1 | 3 - 4 | 386 | 71 | ||
NM_127520.4 | NP_179552.1 | 4 - 5 | 662 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
