Assay Details
Gene Symbol: EDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114680.4 | NP_190394.3 | 13 - 14 | 1614 | 84 | |
GenBank mRNA | AY136413.1 | - | 13 - 14 | 1530 | 84 | |
BT012191.1 | - | 13 - 14 | 1473 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
