Assay Details
Gene Symbol: EDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114680.4 | NP_190394.3 | 24 - 25 | 2561 | 88 | |
GenBank mRNA | AY136413.1 | - | 24 - 25 | 2477 | 88 | |
BT012191.1 | - | 24 - 25 | 2420 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
