Assay Details
Gene Symbol: EDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114680.4 | NP_190394.3 | 2 - 3 | 646 | 78 | |
GenBank mRNA | AY136413.1 | - | 2 - 3 | 562 | 78 | |
BT012191.1 | - | 2 - 3 | 505 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
