Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120804.3 | NP_196339.1 | 3 - 4 | 895 | 65 | |
GenBank mRNA | AF428411.1 | - | 3 - 4 | 797 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
