Assay Details
Gene Symbol: RBOHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203321.1 | NP_001190250.1 | 11 - 12 | 2108 | 76 | |
NM_120821.2 | NP_196356.1 | 10 - 11 | 2249 | 76 | ||
GenBank mRNA | BT003857.1 | - | 10 - 11 | 2249 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
