Assay Details
Gene Symbol: ALKBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335786.1 | NP_001325000.1 | 2 - 3 | 531 | 152 | |
NM_001335787.1 | NP_001324995.1 | 3 - 4 | 376 | 152 | ||
NM_001335788.1 | NP_001324999.1 | 3 - 4 | 421 | 152 | ||
NM_001335789.1 | NP_001324998.1 | 3 - 4 | 376 | 152 | ||
NM_001335790.1 | NP_001324997.1 | 3 - 4 | 336 | 152 | ||
NM_001335791.1 | NP_001324996.1 | 2 - 3 | 367 | 152 | ||
NM_001335792.1 | NP_001324994.1 | 3 - 4 | 257 | 152 | ||
NM_127792.4 | NP_565530.1 | 3 - 4 | 376 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
