Assay Details
Gene Symbol: ALKBH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335786.1 | NP_001325000.1 | 6 - 7 | 1178 | 88 | |
NM_001335788.1 | NP_001324999.1 | 7 - 8 | 1068 | 88 | ||
NM_001335789.1 | NP_001324998.1 | 7 - 8 | 1023 | 88 | ||
NM_001335790.1 | NP_001324997.1 | 7 - 8 | 983 | 88 | ||
NM_001335793.1 | NP_001324993.1 | 4 - 5 | 720 | 88 | ||
NM_127792.4 | NP_565530.1 | 7 - 8 | 1023 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
