Assay Details
Gene Symbol: MES7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127920.3 | NP_179937.1 | 1 - 2 | 536 | 75 | |
GenBank mRNA | BT010709.1 | - | 1 - 2 | 475 | 75 | |
BT012414.1 | - | 1 - 2 | 375 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
