Assay Details
Gene Symbol: ACAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124198.4 | NP_568694.2 | 7 - 8 | 949 | 69 | |
NM_180823.3 | NP_851154.1 | 6 - 7 | 1019 | 69 | ||
GenBank mRNA | AF364059.1 | - | 7 - 8 | 818 | 69 | |
AK317237.1 | - | 7 - 8 | 722 | 69 | ||
AY059736.1 | - | 5 - 6 | 687 | 69 | ||
AY091271.1 | - | 5 - 6 | 616 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
