Assay Details
Gene Symbol: BAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345522.1 | NP_001332748.1 | 2 - 3 | 713 | 112 | |
NM_001345523.1 | NP_001332747.1 | 2 - 3 | 660 | 112 | ||
NM_125603.3 | NP_568950.2 | 2 - 3 | 713 | 112 | ||
NM_180915.1 | NP_851246.1 | 2 - 3 | 520 | 112 | ||
GenBank mRNA | AK228933.1 | - | 2 - 3 | 494 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
